Rabbit polyclonal antibody to Bcl-2. 1 * 100 µl
Catalog # ANTIA81033-100
Supplier: Antibodies.com
New Product
Rabbit polyclonal antibody to Bcl-2. 1 * 100 µl
Catalog # ANTIA81033-100
Supplier: Antibodies.com
New Product
Frequently Bought Together
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Supplier: BIOCISION
Storage plates, 96-well, MASTERBLOCK®
Supplier: Greiner Bio-One
Cell culture multiwell plates, CELLSTAR®
Supplier: Greiner Bio-One
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
VWR®, Sample Dippers
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Centrifuge Tubes, Conical Bottom, 5 ml
Supplier: VWR Collection
VWR®, Microcentrifuge Tubes with Socket Screw Caps
Supplier: VWR Collection
Individually Wrapped Serological Pipettes, Falcon®
Supplier: Corning
VWR®, Centrifuge Tubes, Light-Sensitive
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR®, Centrifuge Tubes, Freestanding, with Flat Screw Caps
Supplier: VWR Collection
Cell culture flasks with standard or filter caps, CELLSTAR®
Supplier: Greiner Bio-One
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
BOTTLE PET 250ML SQUARE T/E GREEN CAP 1 * 200 items
Supplier: LAB3 OWN PRODUCTION
Gel blotting sheets, GB005, Whatman™
Supplier: Whatman products (Cytiva)
Storage tubes, 2D barcoded, Matrix™
Supplier: THERMO MATRIX TECHNOLOGIES