SYNTHETIC NANODISC HUMAN E-SYT1 PROT 1 * 50 µG
Catalog # ANTIA317341-50
Supplier: Antibodies.com
New Product
SYNTHETIC NANODISC HUMAN E-SYT1 PROT 1 * 50 µG
Catalog # ANTIA317341-50
Supplier: Antibodies.com
New Product
Frequently Bought Together
Bottles, narrow neck, octagonal, PET, with screw caps, Corning®
Supplier: Corning
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR®, Transzfer pipetták, Standard Line
Supplier: VWR Collection
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Tárolódobozok fagyasztócsövekhez, karton, 136x136 mm
Supplier: VWR Collection
Deep well and assay plates, Axygen®
Supplier: Corning
Centrifuge tubes, 15 ml, Corning®
Supplier: Corning
Eppendorf® Protein LoBind® mikrocentrifuga csövek
Supplier: EPPENDORF
Cryoboxes, B69/B50/B80/B99-Series
Supplier: GLW
VWR®, Cellulóz dugók
Supplier: VWR Collection
VWR®, Tárolódobozok fagyasztócsövekhez, karton, 133×133 mm
Supplier: VWR Collection
VWR®, Cryoboxes with Dividers
Supplier: VWR Collection
Szűrőplatek, 384 mintahelyes, AcroPrep™, 100 µl
Supplier: Cytiva (Formerly Pall Lab)
[EN]PARTITION INSERTS FOR CRYOBOX 10X10 1 * 10 DB
Supplier: Roth Carl
Strips of 8 PCR tubes with attached caps, 0,2 ml, Amplitube™
Supplier: Simport Scientific