Recombinant rabbit monoclonal [RM371] antibody to CD163. 1 * 50 µl
ANTIA121491-50
New Product
Recombinant rabbit monoclonal [RM371] antibody to CD163. 1 * 50 µl
ANTIA121491-50
New Product
Frequently Bought Together
PCR tubes and caps, Axygen®
: Corning
Deep well plates and cover mats
: Brand
VWR® Premium Line, Serological Pipettes
: VWR Collection
VWR®, Cryoboxes, Half Size, Cardboard
: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
Serological pipettes, Stripette™
: Corning
Plates and modules, FluoroNunc™ and LumiNunc™
: Thermo Fisher Scientific
Media bottle closures, Top Works™ system, Nalgene®
: Thermo Fisher Scientific
Microplates, 384-well, non binding
: Greiner Bio-One
Accessories and consumables for KingFisher™ Flex
: Thermo Fisher Scientific
Microplate Self-adhesive Film and Foil
: EPPENDORF
Storage tubes, 2D barcoded, Matrix™
: THERMO MATRIX TECHNOLOGIES
Sampling spoons, sterile, Sterileware®, SP Bel-Art
: Bel-Art Products, a Part of SP