Recombinant rabbit monoclonal [RM331] antibody to PAX5. 1 * 50 µl
Catalog # ANTIA121382-50
Supplier: Antibodies.com
New Product
Recombinant rabbit monoclonal [RM331] antibody to PAX5. 1 * 50 µl
Catalog # ANTIA121382-50
Supplier: Antibodies.com
New Product
Frequently Bought Together
Solid black and white polystyrene microplates, 384-well, Corning®
Supplier: Corning
VWR®, Strips of PCR Tubes and Caps
Supplier: VWR Collection
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
Supplier: VWR Collection
Cryoboxes, PS, CryoLine™, Nunc™
Supplier: Thermo Fisher Scientific
PCR plates, 384-well, Thermo-Fast® 384
Supplier: Thermo Fisher Scientific
VWR®, Immunoassay Microplates and Strips
Supplier: VWR Collection
Culture tubes, sterile
Supplier: Simport Scientific
Microcentrifuge tubes, 0,5/1,5/1,8 ml, Micrewtube®
Supplier: Simport Scientific
Microcentrifuge tubes, 0,5/1,5/2,0 ml, with/without screw caps
Supplier: Thermo Fisher Scientific
Microcentrifuge tubes, 0,5/1,5/2,0 ml
Supplier: RATIOLAB
Cryoboxes
Supplier: SARSTEDT
Storage tubes, 2D barcoded, Matrix™
Supplier: THERMO MATRIX TECHNOLOGIES
Microcentrifuge tubes, 0,5/1,5/1,8 ml, Micrewtube®
Supplier: Simport Scientific
Inoculating loops and needles, Ino-Loop™
Supplier: Simport Scientific
Microtube Storage Boxes
Supplier: HEATHROW