0 Ft
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Hungary
ContactUsLinkComponent
Recombinant rabbit monoclonal [RM126] antibody to Kappa Light Chain. 1 * 50 µG
ANTIA121308-50
:
Antibodies.com
New Product
Recombinant rabbit monoclonal [RM126] antibody to Kappa Light Chain. 1 * 50 µG
ANTIA121308-50
:
Antibodies.com
New Product
Frequently Bought Together
VWR® Multiwell Cell Culture Plates
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
Cell strainers, Corning®
: Corning
VWR® Standard Line Transfer Pipettes
: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
VWR®, Ice Pans, with or without Lid
: VWR Collection
Serological pipettes, Stripette™
: Corning
Cryoboxes, B69/B50/B80/B99-Series
: GLW
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
Petri dishes
: Greiner Bio-One
Eppendorf® Protein LoBind® Microcentrifuge Tubes
: EPPENDORF
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
Filter plates, 384-well, 100 µl, AcroPrep™
: Cytiva (Formerly Pall Lab)