ANTI-PMPCA / INPP5E ANTIBODY 1 * 100 µl
ANTIA16385-100
New Product
Frequently Bought Together
Corning® Microplate fedők
: Corning
Színes betétek fagyaszócsövekhez, CryoColorCode, Nunc™
: Thermo Fisher Scientific
VWR®, Tárolódobozok fagyasztócsövekhez, karton, 133×133 mm
: VWR Collection
Eppendorf® DNA LoBind Plates
: EPPENDORF
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
: VWR Collection
Cell scrapers
: Greiner Bio-One
Storage tubes, 2D barcoded, Matrix™
: THERMO MATRIX TECHNOLOGIES
PCR microtubes without caps, 0,2 ml, Amplitube™
: Simport Scientific
VWR®, Microcentrifuge Tubes with Socket Screw Caps
: VWR Collection
Centrifuga csövek, kerek fenekű, PP
: KARTELL
Screw caps for T501 sample tubes
: Simport Scientific
Cell culture plates and Multidishes, Nunclon™ Sphera™
: Thermo Fisher Scientific