Rabbit polyclonal antibody to MSH6. 1 * 100 µl
Catalog # ANTIA14392-100
Supplier: Antibodies.com
New Product
Rabbit polyclonal antibody to MSH6. 1 * 100 µl
Catalog # ANTIA14392-100
Supplier: Antibodies.com
New Product
Frequently Bought Together
Serological pipettes, Stripette™
Supplier: Corning
TISSUE CULT PLATE 24/1ML FLAT BOTTOM RED 1 * 50 items
Supplier: SARSTEDT
96-well solid black and white polystyrene microplates, Corning®
Supplier: Corning
Caps for sample tubes, tamper evident
Supplier: Simport Scientific
Scoops, disposable, PharmaScoop®, SteriWare®
Supplier: Sampling Systems
Detectable scoops, SteriPlast®
Supplier: BURKLE
Screw caps for T501 sample tubes
Supplier: Simport Scientific
Storage tubes, 2D barcoded, Matrix™
Supplier: THERMO MATRIX TECHNOLOGIES
Cryoboxes
Supplier: SARSTEDT
VWR®, PCR/qPCR Plates, 96-Wells, Plastic
Supplier: VWR Collection
Measuring scoops
Supplier: KARTELL
Labtop coolers, Nalgene®
Supplier: Thermo Fisher Scientific
Petri dishes, square
Supplier: Anicrin
Insert System, BRAND<i>plates</i>®
Supplier: Brand
VWR®, Cell Culture Insert Plates, PET
Supplier: VWR Collection