BACKGROUND BLOCKER (1X) 1 * 100 mL
Catalog # ANTIA319555-100
Supplier: Antibodies.com
New Product
Frequently Bought Together
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Microcentrifuge Tubes, Low-Binding
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR® Blender Bags
Supplier: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
Containers with snap caps, Corning® Gosselin™
Supplier: Corning
Multiple well cell culture plates, Costar®
Supplier: Corning