MOUSE HRP DETECTION REAGENT 1 * 100 T
Catalog # ANTIA319539-100
Supplier: Antibodies.com
New Product
Frequently Bought Together
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
Serological pipettes, Stripette™
Supplier: Corning
Eppendorf® DNA LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Centrifuge Tubes, High Performance, Flat or Plug Cap, PP
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR®, Cell Culture Flasks, Non Treated or Treated
Supplier: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF