Skip to Footer
Bővített
Bővített
SDS
Tanúsítványok
Gyártói adatok
0
0 Ft
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Hungary
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
8mm Combination Seal: PP Screw Cap, black, centre hole, 8-425 thread Silicone white/PTFE red, 45 shore A, 1.3mm, slitted (ECO-Line) 1 * 2.000 items
Catalog # LAPH08153464
Supplier:
LA PHA PACK
Expand image
8mm Combination Seal: PP Screw Cap, black, centre hole, 8-425 thread Silicone white/PTFE red, 45 shore A, 1.3mm, slitted (ECO-Line) 1 * 2.000 items
Catalog # LAPH08153464
Supplier:
LA PHA PACK
Frequently Bought Together
Aluminium oxide activated basic for chromatography
Supplier: VWR Chemicals
Avantor® ACE® C18, HPLC Columns, 5 µm
Supplier: Avantor
VWR®, Screw Neck Headspace Vials ND18
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, TLC Aluminium Plates, Silica, Unmodified Layers
Supplier: VWR Collection
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
VWR®, Screw Caps for Screw Neck Vials ND24
Supplier: VWR Collection
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Snap Cap Vials
Supplier: VWR Collection
Exhaust filters, V3.0
Supplier: S.C.A.T.
Florisil® 60-100 mesh for chromatography
Supplier: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection