Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 Ft
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Hungary
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
7 COMPONENTS: 2000MG/L EACH 1 * 5 Ampoul
Catalog # CMPLRD0615111.5
Supplier:
CUSTOM MADE CHEMICALS LAB
Expand image
7 COMPONENTS: 2000MG/L EACH 1 * 5 Ampoul
Catalog # CMPLRD0615111.5
Supplier:
CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Sodium hypochlorite (12% Cl₂) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
UltraClear® for histology, J.T.Baker®
Supplier: Avantor
Ammonium formate ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
VWR® Cryolab Q Path, Freezing Aerosol
Supplier: VWR Chemicals
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
Test kits, photometric, ammonium, LCK cuvette tests
Supplier: Hach
Test kits, photometric, Nitrate, LCK cuvette tests
Supplier: Hach