Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 Ft
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Hungary
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
COMPONENTS MIX 5 1 * 5 Ampoul
CMPLRD1087381.5
:
CUSTOM MADE CHEMICALS LAB
COMPONENTS MIX 5 1 * 5 Ampoul
CMPLRD1087381.5
:
CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
: VWR Chemicals
Potassium hydroxide 85.0-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
: VWR Chemicals
Test kits, photometric, COD, LCK cuvette tests
: Hach