IL-21 (HUMAN), (RECOMBINANT) 1 * 100 µG
Catalog # ENZOENZPRT2480100
Supplier: Enzo Life Sciences
Frequently Bought Together
Transfer pipettes
Supplier: RATIOLAB
MicroWell™ Plates, 96 well, Nunc-Immuno™
Supplier: Thermo Fisher Scientific
VWR®, Strips of PCR Tubes and Caps
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Cell culture flasks, Corning®
Supplier: Corning
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Cryoboxes, 133×133 mm
Supplier: RATIOLAB
Serological pipettes, Stripette™
Supplier: Corning
Eppendorf® DNA LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR® Blender Bags
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
VWR®, Petri Dish, PS
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Containers with snap caps, Corning® Gosselin™
Supplier: Corning
Multiple well cell culture plates, Costar®
Supplier: Corning