SONIDEGIB DIPHOSPHATE 1 * 10 mg
Catalog # ENZOENZCHM2370010
Supplier: Enzo Life Sciences
Frequently Bought Together
Potassium hexacyanoferrate(II) trihydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Potassium nitrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium fluoride 99.0-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Sodium chloride ≥98%, TECHNICAL
Supplier: VWR Chemicals
Sodium sulphite 95.0-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Silver nitrate 99.8-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Barium chloride dihydrate ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Sodium sulphate, anhydrous 98.5-101.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals