Skip to Header
Skip to Main Content
Skip to Footer
Bővített
Bővített
SDS
Tanúsítványok
Gyártói adatok
0
0 Ft
Order Entry
Login
/
Regisztráció
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Hungary
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
1 COMPONENT C8H5O4K 30MG/L IN H2O 1 * 500 mL
Catalog # CMPLRM107229L5
Supplier:
CUSTOM MADE CHEMICALS LAB
Expand image
1 COMPONENT C8H5O4K 30MG/L IN H2O 1 * 500 mL
Catalog # CMPLRM107229L5
Supplier:
CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium hydroxide 32% in aqueous solution, GPR RECTAPUR®, pure for nitrogen analysis
Supplier: VWR Chemicals
1-Octanesulphonic acid sodium salt ≥98%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Test strips, nitrate/nitrite, Dosatest
Supplier: VWR Chemicals
Sodium hydroxide 35% in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals