0 Ft
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Hungary
ContactUsLinkComponent
1 COMPONENT C8H5O4K 5000 MG/L IN H2O 1 * 500 mL
CMPLRM005711L5
:
CUSTOM MADE CHEMICALS LAB
1 COMPONENT C8H5O4K 5000 MG/L IN H2O 1 * 500 mL
CMPLRM005711L5
:
CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Paraffin, Solidification point: 54 - 56 °C, Q Path® for microscopy
: VWR Chemicals
Test strips, peroxide, Dosatest
: VWR Chemicals
Reagents for chlorine determination using Lovibond® MD photometers
: Lovibond
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
: VWR Chemicals
VWR® Rota®, pH Indicator Paper, 1 to 14
: VWR Chemicals
Test kits, photometric, COD, LCK cuvette tests
: Hach