The MB Fit24™ cfDNA Kit CE IVD is designed for rapid and reliable isolation of circulating DNA from up to 4 mL plasma/serum samples with the use of the MagBinder® Fit24 Nucleic Acid Purification System. 1 * 48 items
Catalog # OMEGB3298-10-48PFC
Supplier: OMEGA BIO-TEK
Frequently Bought Together