0
0 Ft
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Hungary
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
PEEK-KAPILLARE 1/16INAD 0.18MM ID YELLOW max 400bar gelb Farbstreifen-markiert wähbare Länge, Preis/Meter 1 * 1 items
Catalog # KNAUA2523
Supplier:
KNAUER
Expand image
PEEK-KAPILLARE 1/16INAD 0.18MM ID YELLOW max 400bar gelb Farbstreifen-markiert wähbare Länge, Preis/Meter 1 * 1 items
Catalog # KNAUA2523
Supplier:
KNAUER
Frequently Bought Together
Accessories for multi channel pipettes, mechanical, Research® plus
Supplier: EPPENDORF
Accessories for Pipet-Aid® controllers
Supplier: DRUMMOND
Installation of autoclave >100L at customer site. The device must be at the place of use. Functionality test and users short functi ons introduction. Travel fees included (cost for air travel excluded). 1 * 1 Each
Supplier: AVANTOR SERVICES
Accessories for DOSE IT
Supplier: Integra