Order Entry
Hungary
ContactUsLinkComponent
The Ig-kappa/Ig-lambda (Human) CISH Kit is intended to be used for the simultaneous qualitative detection of human Ig-kappa and Ig-l ambda light chain mRNA in formalin-fixed, paraffin-embedded specimens, such as lymphomas, by CISH. 1 * 1 KIT
Catalog # ABNOKA6908.1KIT
Supplier:  Abnova
The Ig-kappa/Ig-lambda (Human) CISH Kit is intended to be used for the simultaneous qualitative detection of human Ig-kappa and Ig-l ambda light chain mRNA in formalin-fixed, paraffin-embedded specimens, such as lymphomas, by CISH. 1 * 1 KIT
Catalog # ABNOKA6908.1KIT
Supplier:  Abnova
Frequently Bought Together