0 Ft
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Hungary
ContactUsLinkComponent
1 component: cis-1,2-Dichloroethene [CAS:156-59-2] 2000mg/l in Methanol 1 * 5 Ampoul
Catalog # CMPLRD0100981.5
Supplier:
CUSTOM MADE CHEMICALS LAB
Expand image
1 component: cis-1,2-Dichloroethene [CAS:156-59-2] 2000mg/l in Methanol 1 * 5 Ampoul
Catalog # CMPLRD0100981.5
Supplier:
CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
UltraClear® for histology, J.T.Baker®
Supplier: Avantor
Sodium hydroxide ≥98%, pellets, GPR RECTAPUR®
Supplier: VWR Chemicals
Paraffin, Solidification point: 54 - 56 °C, Q Path® for microscopy
Supplier: VWR Chemicals
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium hypochlorite (3,5% Cl₂) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Reagents for copper determination using Lovibond® MD photometers
Supplier: Lovibond