Order Entry
Hungary
ContactUsLinkComponent
1 component: Perfluorohexanesulfonic acid [CAS:355-46-4] 100mg/l in Methanol CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store in a refrigerator at temperatures between 2C to 8C 1 * 1 mL
Catalog # CMPLRD0153081
1 component: Perfluorohexanesulfonic acid [CAS:355-46-4] 100mg/l in Methanol CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store in a refrigerator at temperatures between 2C to 8C 1 * 1 mL
Catalog # CMPLRD0153081
Frequently Bought Together