PRECISION HYDROGEN TRACE 500CC AGILENT 1 * 1 items
Katalógus# PEAK74-0500
Supplier: PEAK SCIENTIFIC
Frequently Bought Together
VWR® Counter-Pen™ Cell Counter
Supplier: VWR Collection
Accessories for OxiTop® and OxiTop® Control
Supplier: WTW
Accessories for VWR® V-1200, UV-1600PC, UV-3100PC
Supplier: COLLECTION PREFIX HPLC
VWR®, Portable Turbidity Meter
Supplier: VWR Collection
Accessories for Mettler balances
Supplier: Mettler - Toledo
Accessory for airflow indicator
Supplier: Dräger
Titration beakers
Supplier: Mettler - Toledo
VWR® PH 20 pH Meter, Handheld
Supplier: VWR Collection
Conductivity standards
Supplier: WTW
Accessories for cell counter, EVE™
Supplier: NANOENTEK
VWR® pH CO 1030, pH/Conductivity Tester
Supplier: VWR Collection
VWR®, Hand Tally Counter
Supplier: VWR Collection
pH/mV/°C meters, bench, inoLab® pH 7110 / 7310
Supplier: WTW
Plastic cuvette for LICO
Supplier: Hach
Melting point and boiling point capillaries and tubes
Supplier: BUCHI