2,6-DIBROMO-4-METHYLANISOLE 1 * 5 g
Catalog # APOSOR900776-5G
Supplier: Apollo Scientific
Frequently Bought Together
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals
Methanol, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Toluene ≥99.5% ACS
Supplier: Thermo Fisher Scientific
Diisopropyl ether ≥98.5% (by GC) stabilised, Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
Petroleum spirit, 40…65 °C, Technical Grade
Supplier: Thermo Fisher Scientific
Acetone, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Toluene ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
1-Methylpyrrolidine 98%
Supplier: Thermo Fisher Scientific
2-Nitrophenylhydrazine, moistened 30% with water 97%
Supplier: Thermo Fisher Scientific
Toluene, extra dry 99.85%, AcroSeal®
Supplier: Thermo Fisher Scientific
Glycerine ≥83.5-88.5% (w/w) in aqueous solution, pure
Supplier: Thermo Fisher Scientific
ALCH, CHROMASOLV™ for HPLC (denatured with 5% isopropanol and 5% methanol), Riedel-de Haën™
Supplier: Honeywell Chemicals
Diethyl ether stabilised, BAKER, J.T.Baker®
Supplier: Avantor
Diethyl ether ≥99% stabilised for HPLC
Supplier: Thermo Fisher Scientific
Polyvinyl alcohol, M.W. ∼ 124,000 99-100%, hydrolysed
Supplier: Thermo Fisher Scientific