3-CHLORO-5-FLUORO-2-METHOXYBENZALDEHYDE 1 * 5 g
Catalog # APOSPC57537-5G
Supplier: Apollo Scientific
3-CHLORO-5-FLUORO-2-METHOXYBENZALDEHYDE 1 * 5 g
Catalog # APOSPC57537-5G
Supplier: Apollo Scientific
Frequently Bought Together
N,N-Dimethylformamide ≥99.9%, SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Maleic acid 99%
Supplier: Thermo Fisher Scientific
Toluene, extra dry over Molecular Sieve 99.85%, AcroSeal®
Supplier: Thermo Fisher Scientific
Ethanol denatured ≥96%, meets analytical specification of Ph.Eur. (denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex®)
Supplier: Thermo Fisher Scientific
4-Methyl-2-pentanone, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
tert-Butanol, anhydrous ≥99.5%
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.5% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
N,N-Dimethylacetamide, extra dry 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Cyclohexane, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetone ≥99.0% Ph. Eur., Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Methanol ≥99.6%, Laboratory Reagent
Supplier: Honeywell Chemicals