ISOSORBIDE-2-MONONITRATE 5-O-?-D-GLUCURO 1 * 5 mg
Catalog # APOSBICL2321-5MG
Supplier: Apollo Scientific
ISOSORBIDE-2-MONONITRATE 5-O-?-D-GLUCURO 1 * 5 mg
Catalog # APOSBICL2321-5MG
Supplier: Apollo Scientific
Frequently Bought Together
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
VWR®, Ice Pans, with or without Lid
Supplier: VWR Collection
Eppendorf® DNA LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
Dimethyl sulphoxide ≥99.5%, BioChemica® cell culture reagent
Supplier: PanReac AppliChem
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
96 MicroWell™ plates, Nunclon™∆
Supplier: Thermo Fisher Scientific
Adhesive PCR film and foil seals
Supplier: Thermo Fisher Scientific
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
Supplier: VWR Chemicals
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
Cryogenic tubes, CryoTubes™, Nunc™
Supplier: Thermo Fisher Scientific
Methanol ≥99.90%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals