Order Entry
Hungary
ContactUsLinkComponent
LABOWATER DM 4000 Ion exchanger cartridge, Pressure-resistant made of stainless steel Volume: 26,5 l Capacity at 10 GSG: 4200 l Dimensions: 239 x 703 mm (diam x H) 1 * 1 items
Catalog # SHPS812400000000
Supplier:  SHP Steriltechnik
LABOWATER DM 4000 Ion exchanger cartridge, Pressure-resistant made of stainless steel Volume: 26,5 l Capacity at 10 GSG: 4200 l Dimensions: 239 x 703 mm (diam x H) 1 * 1 items
Catalog # SHPS812400000000
Supplier:  SHP Steriltechnik
Frequently Bought Together