Order Entry
Hungary
ContactUsLinkComponent
Manual Pinch Valve, 5/32" to 1/4" tube OD
Manual Pinch Valve, 5/32" to 1/4" tube OD
Catalog # MFLX98002-00
undefined
Manual Pinch Valve, 5/32" to 1/4" tube OD
Catalog # MFLX98002-00
About this item
An ideal option to secure tubing.
  • Constructed of durable Delrin®
Fluid contacts only the tubing—ideal for use in pharmaceutical and food industries where eliminating contamination is critical. Obtain precise and repeatable flow rates using easy-to-read metered scale on valve body. Valves are made of Delrin®. Tubing is not included—order flexible tubing with durometer (Shore A) of 60 or less from our "Tubing" section.
  • Connection Type:
    Pinch Valve for Tubing
  • Description:
    Manual Pinch Valve, 5/32" to 1/4" tube OD
  • Environmentally Preferable:
  • Pinch valve tube size:
    5/32" to 1/4" OD
  • Warranty:
    3 Months
  • Pk:
    1
Frequently Bought Together