Mounting Bracket Kit for Liquid/Solids In-Line Separators, 1
Catalog # MFLX29995-52
Supplier: Avantor Fluid Handling
About this item
Optimize performance of your Liquid-Solids Separator.
- Keep your Separators running properly with these kits
Choose from a wide range of replacement parts and accessories to ensure top performance of your Liquid-Solids Separator / Concentrator Units. Purge kits are available in manual or automatic forms so solids may be purged or bled as needed.
Mounting bracket kits are designed to support the separator at a near-upright position.
- Description:Mounting Bracket Kit for Liquid/Solids In-Line Separators, 1
- Environmentally Preferable:
- Warranty:1 year
- Compatible with:29995-06, 29995-08, 29995-10
- Pk:1
Frequently Bought Together