Mounting Bracket Kit for In-Line Separators, 1/2" to 3/4" Inlet/Outlet
Catalog # MFLX29995-50
Gyártó: Avantor Fluid Handling
Termék információk
Optimize performance of your Liquid-Solids Separator.
- Keep your Separators running properly with these kits
Choose from a wide range of replacement parts and accessories to ensure top performance of your Liquid-Solids Separator / Concentrator Units. Purge kits are available in manual or automatic forms so solids may be purged or bled as needed.
- Leírás:Mounting Bracket Kit for Liquid/Solids In-Line Separators, 1/2" to 3/4" Inlet/Outlet
- Environmentally Preferable:
- Garancia:1 year
- Kompatibilis:29995-00, 29995-02, 29995-04
- Kiszerelés:1
Frequently Bought Together