Order Entry
Hungary
Orders LinkContactUsLinkComponent
VWR®, Inserts for Shell Vials

VWR®, Inserts for Shell Vials

Micro-insert, 13 mm top, 0,10 ml, Conical bottom, 5×34 mm, Borosilicate glass 5.1, Clear, No
Catalog # 548-0044A
Supplier:   VWR Collection
VWR®, Inserts for Shell Vials
Catalog # 548-0044A
Supplier:  VWR Collection
VWR®, Inserts for Shell Vials
VWR®, Inserts for Shell Vials

Micro-insert, 13 mm top, 0,10 ml, Conical bottom, 5×34 mm, Borosilicate glass 5.1, Clear, No

Price
171 000 Ft
Log in for availability
1 * 1.000 DB
Product Details & Documents

Documents

Specifications
  • Cap included:
    No
  • Ø×H:
    5×34 mm
  • Capacity:
    0,10 ml
  • Colour:
    Clear
  • Description:
    Micro-insert, 13 mm top
  • Material:
    Borosilicate glass 5.1
  • Seq:
    01
  • Base:
    Conical bottom