HIS MAG SEPHAROSE EXCEL 100ML (10%) 1 * 100 mL
Katalógus# 29-3583-45
Supplier: Cytiva
Frequently Bought Together
Microspatulas, LaboPlast® / SteriPlast®
Supplier: BURKLE
VWR®, Flat Screw Caps for Microcentrifuge Tubes
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
Individually Wrapped Serological Pipettes, Falcon®
Supplier: Corning
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Test tubes
Supplier: Greiner Bio-One
Cell scrapers and cell lifters
Supplier: VWR Collection
Reagent/centrifuge tube with screw cap
Supplier: Greiner Bio-One
Vial, flip-top
Supplier: Capitol Europe
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
96-Well cell culture plates, Falcon®
Supplier: Corning
Scoops
Supplier: VITLAB
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR® Bottles, Water Sampling
Supplier: VWR Collection