Order Entry
Hungary
Orders LinkContactUsLinkComponent
VWR®, Cold Filter Plugging Point (CFPP) standards, Certified Reference Material

VWR®, Cold Filter Plugging Point (CFPP) standards, Certified Reference Material

CFPP Standard (CRM), –21,7 °C, Aluminium bottle, 250 mL
Catalog # 99102.230
Supplier:   VWR Chemicals
VWR®, Cold Filter Plugging Point (CFPP) standards, Certified Reference Material
Catalog # 99102.230
Supplier:  VWR Chemicals
CAS Number:  
VWR®, Cold Filter Plugging Point (CFPP) standards, Certified Reference Material
VWR®, Cold Filter Plugging Point (CFPP) standards, Certified Reference Material

CFPP Standard (CRM), –21,7 °C, Aluminium bottle, 250 mL

CAS Number:  
Price
129 000 Ft
Log in for availability
1 * 250 mL
Product Details & Documents

Cold Filter Plugging Point (CFPP) Certified Reference Materials are for the verification of laboratory instruments used in testing petroleum and derivative products for Cold Filter Plugging Point (CFPP) according to IP 309, EN 116 and ASTM D6371 also other equivalent test methodology.

  • Tested in accordance with ASTM / EN or IP test method protocols
  • Low level of uncertainty achieves maximum data accuracy
  • Supplied in tamper-evident UN compliant security pack with 12 month shelf life

Certifications: Fully certified in accordance with ISO 17034 under accreditation by UKAS.

Specifications

Specifications

  • Pack type:
    Aluminium bottle
  • Pk:
    250 mL
  • Description:
    CFPP Standard (CRM)
  • Temperature:
    –21,7 °C

Specifications