Order Entry
Hungary
ContactUsLinkComponent
Aluminiumfolie, weich, glatt, 0,020 x 1.000 mm, 150 lfm/Rolle 1 * 1 Roll
Catalog # KORF16297
Supplier:  Korff
Aluminiumfolie, weich, glatt, 0,020 x 1.000 mm, 150 lfm/Rolle 1 * 1 Roll
Catalog # KORF16297
Supplier:  Korff
Frequently Bought Together