Order Entry
Hungary
ContactUsLinkComponent
Pharmaceutical Refrigerator. 684 litre (24.2 cu.ft) interior capacity. Microprocessor temperature control (range 2C to 23C) with LED display to 0.1C and audible & visual high/low temperature alarms and door open alarm. Power failure alarm and remo 1 * 1 items
Catalog # SANYMPR-722
Supplier:  PHC Corporation
Pharmaceutical Refrigerator. 684 litre (24.2 cu.ft) interior capacity. Microprocessor temperature control (range 2C to 23C) with LED display to 0.1C and audible & visual high/low temperature alarms and door open alarm. Power failure alarm and remo 1 * 1 items
Catalog # SANYMPR-722
Supplier:  PHC Corporation
Frequently Bought Together