Order Entry
Hungary
Orders LinkContactUsLinkComponent
 

VWR® ARISTAR®, Multi-Element Standards, Anionic for Ion Chromatography

IC multi-element standard solution V, 10 - 200 mg/L, H₂O, F, Br, NO₃, PO₄, Cl, SO₄, in water, 100 mL
Catalog # 87040.180
Supplier:   VWR Chemicals
VWR® ARISTAR®, Multi-Element Standards, Anionic for Ion Chromatography
Catalog # 87040.180
Supplier:  VWR Chemicals
CAS Number:  
VWR® ARISTAR®, Multi-Element Standards, Anionic for Ion Chromatography
VWR® ARISTAR®, Multi-Element Standards, Anionic for Ion Chromatography

IC multi-element standard solution V, 10 - 200 mg/L, H₂O, F, Br, NO₃, PO₄, Cl, SO₄, in water, 100 mL

CAS Number:  
Price
82 100 Ft
Log in for availability
1 * 100 mL
Specifications

Specifications

  • Concentration:
    10 - 200 mg/L
  • Matrix:
    H₂O
  • Pk:
    100 mL
  • Description:
    IC multi-element standard solution V
  • Contents:
    F, Br, NO₃, PO₄, Cl, SO₄, in water

Specifications