VWR® ARISTAR®, Multi-Element Standards, Anionic for Ion Chromatography
Ion chromatography anion multi-element standard solution II, 1000 mg/L, H₂O, Cl, NO₃, SO₄ in water, 500 mL
Supplier: VWR Chemicals
VWR® ARISTAR®, Multi-Element Standards, Anionic for Ion Chromatography
Catalog # 87038.260
Supplier: VWR Chemicals
CAS Number:
Jump to:
Product Details & Documents
- Traceable to NIST
- Manufactured and tested in ISO Guide 34 / ISO 17025 accredited facilities
Delivery information: Supplied with a Certificate of Analysis.
Documents
Certificates
Search by Lot NumberSDS_EN_CH_VWRC87038 SDS_EN_DE_VWRC87038 SDS_EN_NL_VWRC87038 SDS_EN_PL_VWRC87039 SDS_EN_PL_VWRC87039.pdf
SDS
More
spec_Ion Chromatography Anion Multi-element Standard Solution I VWR.pdf spec_Ion Chromatography Anion Multi-element Standard Solution II VWR.pdf spec_Ion Chromatography Multi-element Standard Solution I VWR spec_Ion Chromatography Multi-element Standard Solution V VWR.pdf
Product specification
VWR® for Calibrations
Product Literature
Specifications
Specifications
- Concentration:1000 mg/L
- Matrix:H₂O
- Pk:500 mL
- Description:Ion chromatography anion multi-element standard solution II
- Contents:Cl, NO₃, SO₄ in water
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



