Order Entry
Hungary
ContactUsLinkComponent
Piercing thermometer with alarm, -50...+300:0,1C/1C, max/min function, hold-function, piercing probe of stainless steel 145x3,5mm, with handle, cable PVC c.1000mm, splashproof 1 * 1 items
Catalog # AMARE906200
Supplier:  Amarell
Piercing thermometer with alarm, -50...+300:0,1C/1C, max/min function, hold-function, piercing probe of stainless steel 145x3,5mm, with handle, cable PVC c.1000mm, splashproof 1 * 1 items
Catalog # AMARE906200
Supplier:  Amarell
Frequently Bought Together