Order Entry
Hungary
ContactUsLinkComponent
Cellulose Extraction Thimbles ET/C 25x60 mm 1 * 25 items
Catalog # BINZ142028
Supplier:  Ahlstrom-Munksjö
Cellulose Extraction Thimbles ET/C 25x60 mm 1 * 25 items
Catalog # BINZ142028
Supplier:  Ahlstrom-Munksjö
Frequently Bought Together