BRUSH SOFT BLUE KIT 3 DIAM 2,5,6MM 1 * 1 KIT
Catalog # BSNC53623
Supplier: Basan
Frequently Bought Together
Cryogenic markers, SCIENCE-MARKER™
Supplier: GA
pH buffer solutions
Supplier: Mettler - Toledo
VWR® Nitrile Extra Light, Nitrile Gloves
Supplier: VWR Collection
Colour Lab Tapes
Supplier: GA
VWR®, Laboratory Markers
Supplier: VWR Collection
Labelling tapes, temperature resistant
Supplier: PRECISION DYNAMICS TIMEM
VWR® Basic, Shoe Covers, Anti-Skid
Supplier: VWR Collection
VWR®, Cleanroom Tapes, General Purpose
Supplier: VWR Collection
Laboratory coat, KIMTECH SCIENCE* A7 P+
Supplier: KIMBERLY CLARK
VWR® PolarSafe Dots, Cryo-Labels
Supplier: VWR Collection
VWR®, Visitor Coats, PP, Disposable, Elasticated Wrist
Supplier: VWR Collection
MULTI-SAFE MEDI 6 WITH HANDLES/LABEL 1 * 21 items
Supplier: SARSTEDT
Protective hand creams, Plutect Dual
Supplier: Plum Safety
Cut resistant gloves, HyFlex® 11-425
Supplier: Ansell
Nitrile gloves, Solo 999
Supplier: MAPA
High temperature anti-static cleanroom tapes
Supplier: ULTRATAPE