Angpt2 (Rat) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of rat angiopoietin-2 in cell cult ure supernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5199
Supplier: Abnova
Frequently Bought Together