Order Entry
Hungary
ContactUsLinkComponent
Alcam (Rat) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of rat Alcam in cell culture supern ates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5198
Supplier:  Abnova
Alcam (Rat) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of rat Alcam in cell culture supern ates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5198
Supplier:  Abnova
Frequently Bought Together