Order Entry
Hungary
ContactUsLinkComponent
Tube Vacutainer Plus 6ml 13x100mm serum plastic blue Hemogard closure 1 * 100 items
  BDAM368814
Tube Vacutainer Plus 6ml 13x100mm serum plastic blue Hemogard closure 1 * 100 items
  BDAM368814
Frequently Bought Together