Order Entry
Hungary
ContactUsLinkComponent
Gdf5 (Mouse) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of mouse Gdf5 in cell culture supe rnates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5155
Supplier:  Abnova
Gdf5 (Mouse) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of mouse Gdf5 in cell culture supe rnates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5155
Supplier:  Abnova
Frequently Bought Together