Order Entry
Hungary
ContactUsLinkComponent
CPB2 (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human CPB2 in cell culture supe rnates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5152
Supplier:  Abnova
CPB2 (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human CPB2 in cell culture supe rnates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5152
Supplier:  Abnova
Frequently Bought Together