Order Entry
Hungary
ContactUsLinkComponent
RSPO4 (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human R-Spondin-4 in cell cult ure supernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5146
Supplier:  Abnova
RSPO4 (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human R-Spondin-4 in cell cult ure supernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5146
Supplier:  Abnova
Frequently Bought Together