SFRP5 (HUMAN) ELISA KIT 1 * 1 KIT
Catalog # ABNOKA5139
Supplier: Abnova
Frequently Bought Together
Aceton ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Metanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Víz, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Kesztyűk, nitril
Supplier: VWR Collection
VWR®, Pasteur pipetták, nagy térfogatú
Supplier: VWR Collection
Kesztyűk, nitril, egyszerhasználatos, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR®, szerológiai pipetta, szabvány
Supplier: VWR Collection
VWR®, nitril kesztyű
Supplier: VWR Collection
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Arctörlő kendők
Supplier: VWR Collection