Order Entry
Hungary
ContactUsLinkComponent
Fcgrt (Mouse) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of mouse FCGRT in cell culture su pernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5133
Supplier:  Abnova
Fcgrt (Mouse) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of mouse FCGRT in cell culture su pernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5133
Supplier:  Abnova
Frequently Bought Together