Order Entry
Hungary
ContactUsLinkComponent
Dkk1 (Rat) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of rat DKK1 in cell culture supernat es, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5114
Supplier:  Abnova
Dkk1 (Rat) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of rat DKK1 in cell culture supernat es, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5114
Supplier:  Abnova
Frequently Bought Together