Order Entry
Hungary
ContactUsLinkComponent
Fgf21 (Mouse) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of mouse Fgf21 in cell culture su pernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5092
Supplier:  Abnova
Fgf21 (Mouse) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of mouse Fgf21 in cell culture su pernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5092
Supplier:  Abnova
Frequently Bought Together