Order Entry
Hungary
ContactUsLinkComponent
Storage Boxes for CryoSette Frozen Tissue containers. 40 places. Made of polycarbonate Compatible with leading frozen sample storag e systems. Autoclavable. Temperature range between -196 C and +121 C. Blue. 1 * 5 items
Catalog # SIMPM956-40B
Supplier:  Simport Scientific
Storage Boxes for CryoSette Frozen Tissue containers. 40 places. Made of polycarbonate Compatible with leading frozen sample storag e systems. Autoclavable. Temperature range between -196 C and +121 C. Blue. 1 * 5 items
Catalog # SIMPM956-40B
Supplier:  Simport Scientific
Frequently Bought Together